Merck
  • Intravenous bisphosphonate-related osteonecrosis of the jaw: long-term follow-up of 109 patients.
Intravenous bisphosphonate-related osteonecrosis of the jaw: long-term follow-up of 109 patients.
Oral surgery, oral medicine, oral pathology and oral radiology (2012-10-06)
Amber L Watters, Heidi J Hansen, Tijaana Williams, Joanne F Chou, Elyn Riedel, Jerry Halpern, Steven Tunick, George Bohle, Joseph M Huryn, Cherry L Estilo
ABSTRACT
We report long-term follow-up of patients with intravenous bisphosphonate-related osteonecrosis of the jaw (BRONJ). Medical and dental histories, including type and duration of bisphosphonate treatment and comorbidities, were analyzed and compared with clinical course of 109 patients with BRONJ at Memorial Sloan-Kettering Cancer Center Dental Service. Median onset of BRONJ in months was 21 (zoledronic acid), 30 (pamidronate), and 36 (pamidronate plus zoledronic acid), with a significant difference between the pamidronate plus zoledronic acid and zoledronic acid groups (P = .01; Kruskal-Wallis). The median number of doses for BRONJ onset was significantly less with zoledronic acid (n = 18) than pamidronte plus zoledronic acid (n = 36; P = .001), but not pamidronate alone (n = 29). An association between diabetes (P = .05), decayed-missing-filled teeth (P = .02), and smoking (P = .03) and progression of BRONJ was identified through χ(2) test. This long-term follow-up of BRONJ cases enhances the literature and contributes to the knowledge of BRONJ clinical course.
Sigma-Aldrich
Pamidronate disodium salt hydrate, ≥95% (NMR), solid