Merck
  • Synthesis of polycyclic isoindoline derivatives via tandem Pd-catalyzed coupling, propargyl-allenyl isomerization, [4 + 2] cycloaddition and aromatization reaction.
Synthesis of polycyclic isoindoline derivatives via tandem Pd-catalyzed coupling, propargyl-allenyl isomerization, [4 + 2] cycloaddition and aromatization reaction.
The Journal of organic chemistry (2012-10-12)
Shugao Zhu, Jian Cao, Luling Wu, Xian Huang
ABSTRACT
We report in this paper an interesting sequential reaction involving sequential Sonogashira coupling, propargyl-allenyl isomerization, [4 + 2] cycloaddition and aromatization reaction, which provides a facile method for the synthesis of a variety of polycyclic isoindoline derivatives from easily accessible starting materials.
Sigma-Aldrich
N-Methyl-N-propargylbenzylamine, 97%
Sigma-Aldrich
Pargyline hydrochloride