Merck
  • Consolidation and reconsolidation of memory in black-capped chickadees (Poecile atricapillus).
Consolidation and reconsolidation of memory in black-capped chickadees (Poecile atricapillus).
Behavioral neuroscience (2012-10-17)
Matthew C Barrett, David F Sherry
ABSTRACT
Multiple phases of protein synthesis are necessary for the synaptic modifications that consolidate long-term memory. The reconsolidation hypothesis supposes that information in long-term memory becomes labile and subject to change when retrieved and must be reconsolidated into long-term memory. The current study used the protein synthesis inhibitor anisomycin to examine memory consolidation in birds and to test the reconsolidation hypothesis. Black-capped chickadees store food and usually remember which of their caches they have emptied and which they have left full. In Experiment 1, anisomycin was injected either immediately and 2 hr after food caching, or 4 and 6 hr after food caching. Inhibition of protein synthesis impaired memory for cache sites 24 and 48 hr later. In Experiment 2, it was hypothesized that long-term memory for food caches becomes labile as predicted by the reconsolidation hypothesis when birds search for caches. Anisomycin was administered immediately after chickadees had searched for their caches. Inhibition of protein synthesis should disrupt memory for caches left full if these sites are retrieved from long-term memory and require reconsolidation. Control birds were later more likely to revisit full caches than caches they had emptied. Birds given anisomycin revisited both kinds of caches and did not distinguish between them. This result shows that reconsolidation of full caches into long-term memory is not necessary following search for cache sites, but also shows that protein synthesis-dependent consolidation is required for updating the status of emptied caches.
Sigma-Aldrich
Anisomycin, Ready Made Solution from Streptomyces griseolus, 10 mg/mL in DMSO
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid