Merck
  • Hydrogenation of quinolines, alkenes, and biodiesel by palladium nanoparticles supported on magnesium oxide.
Hydrogenation of quinolines, alkenes, and biodiesel by palladium nanoparticles supported on magnesium oxide.
Dalton transactions (Cambridge, England : 2003) (2012-10-18)
Reena Rahi, Minfeng Fang, Atif Ahmed, Roberto A Sánchez-Delgado
ABSTRACT
A new catalyst composed of Pd nanoparticles supported on MgO has been prepared by the room temperature NaBH(4) reduction of Na(2)PdCl(4) in methanol in the presence of the support. TEM measurements reveal well-dispersed Pd particles of mean diameter 1.7 nm attached to the MgO surface. Further characterization was achieved by ICP-AES, XPS, XRD, H(2) pulse chemisorption and H(2)-TPR. The new catalyst is efficient for the regioselective hydrogenation of the heterocyclic ring of quinolines, as well as for the mild reduction of a variety of alkenes representative of fuel components, and the partial saturation of biodiesel. The new material is considerably more reactive than commercial Pd/SiO(2) and Pd/Al(2)O(3) catalysts under analogous reaction conditions.
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)