Merck
  • Occupational exposure to styrene in the fibreglass reinforced plastic industry: comparison between two different manufacturing processes.
Occupational exposure to styrene in the fibreglass reinforced plastic industry: comparison between two different manufacturing processes.
La Medicina del lavoro (2012-10-20)
Giovanna Tranfo, Monica Gherardi, E Paci, Mariapia Gatto, A Gordiani, Lidia Caporossi, Silvia Capanna, Renata Sisto, B Papaleo, Carla Fiumalbi, Patrizia Garofani
ABSTRACT
Styrene is used in manufacturing fiberglass reinforced plastics: and occupational exposure was related to neurotoxicology and genotoxicity. The sum of the metabolites mandelic and phenylglyoxylic acids is the ACGIH biomarker for occupational exposure with a BEI of 400 mg/g of creatinine in end shift urine corresponding to a airborne styrene concentration of 85 mg/m3. There are two main molding processes, open and closed, the last more effective at controlling worker's styrene exposure. To compare the open molding process to the compression of fiber reinforced resin foils, a kind of closed molding, monitoring the styrene exposure of workers in two production sites (A and B). Environmental Monitoring was carried out by Radiello samplers and Biological Monitoring by means of the determination of MA and PGA with HPLC/MS/MS in 10 workers at Site A and 14 at Site B. The median values for styrene exposure resulted 31.1 mg/m3 for Site A and 24.4 mg/m for Site B, while the medians for the sum of the two metabolites in the end shift urine were 86.7 e 33.8 mg/g creatinine respectively. There is a significant linear correlation between personal styrene exposure and the excretion of styrene metabolites (R = 0.74). As expected the exposure markers of the workers of the two production sites resulted higher in the open process. The analytical results of both environmental and biological monitoring were all below the occupational exposure limits, confirming the efficacy of the protective devices.
Product Description
Sertraline impurity E, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(R)-(−)-Mandelic acid, ChiPros®, produced by BASF, 99%
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
Sigma-Aldrich
Mandelic acid, 99%
Sigma-Aldrich
Phenylglyoxylic acid, purum, ≥98.0% (T)
Sigma-Aldrich
Phenylglyoxylic acid, 97%
Sigma-Aldrich
(R)-(−)-Mandelic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
(R)-(−)-Mandelic acid, 98%
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Filtration Kit, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello Diffusive Bodies, white, configured for general use, pk of 20
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello On-Field Thermometer, Reader, pk of 1
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
radiello Hydrogen Sulfide Starter Kit
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Supelco
Styrene, analytical standard
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Vertical Adapter, threaded for standard use, pk of 20