Merck
  • Combinatorial evaluation of laccase-mediator system in the oxidation of veratryl alcohol.
Combinatorial evaluation of laccase-mediator system in the oxidation of veratryl alcohol.
Biotechnology letters (2012-11-08)
Troy M Larson, Amber M Anderson, Joseph O Rich
ABSTRACT
Laccases play an important role in the biological break down of lignin and have great potential in the deconstruction of lignocellulosic feedstocks. We examined 16 laccases, both commercially prepared and crude extracts, for their ability to oxidize veratryl alcohol in the presence of various solvents and mediators. Screening revealed complete conversion of veratryl alcohol to veratraldehyde catalyzed by a crude preparation of the laccase from Trametes versicolor ATCC 11235 and the mediator TEMPO in 20 % (v/v) tert-butanol.
Sigma-Aldrich
3,4-Dimethoxybenzaldehyde, 99%
Sigma-Aldrich
Veratraldehyde, ≥98%, FG
Sigma-Aldrich
3,4-Dimethoxybenzyl alcohol, 96%