Merck
  • Tuning the reactivity of TEMPO by coordination to a Lewis acid: isolation and reactivity of MCl3(η1-TEMPO) (M = Fe, Al).
Tuning the reactivity of TEMPO by coordination to a Lewis acid: isolation and reactivity of MCl3(η1-TEMPO) (M = Fe, Al).
Journal of the American Chemical Society (2012-11-09)
Jeremiah J Scepaniak, Ashley M Wright, Richard A Lewis, Guang Wu, Trevor W Hayton
ABSTRACT
Addition of 2,2,6,6-tetramethylpiperidine-N-oxyl (TEMPO) to MCl(3) (M = Fe, Al) results in the formation of MCl(3)(η(1)-TEMPO) [M = Fe (1), Al (2)]. Both 1 and 2 oxidize alcohols to generate ketones or aldehydes along with the reduced complexes MCl(3)(η(1)-TEMPOH) [M = Fe (3), Al (4)]. Complexes 1-4 were fully characterized, including analysis by X-ray crystallography. Additionally, control experiments indicated that neither MCl(3) (M = Al, Fe) nor TEMPO are capable of effecting the oxidation of alcohols independently.
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
Aluminum chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Iron(III) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Iron(III) chloride hexahydrate, ACS reagent, 97%
Sigma-Aldrich
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Sigma-Aldrich
TEMPO, 98%
Sigma-Aldrich
TEMPO, purified by sublimation, 99%
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Aluminum chloride hydrate, 99.999% trace metals basis
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran