Merck
  • Ketoacidosis and trace amounts of isopropanol in a chronic alcoholic patient.
Ketoacidosis and trace amounts of isopropanol in a chronic alcoholic patient.
Clinica chimica acta; international journal of clinical chemistry (2012-11-20)
Jessica B Dwyer, Kenichi Tamama
ABSTRACT
Alcohol ketoacidosis is a frequently missed diagnosis, but is well described in the literature. We present a case of ketoacidosis, likely alcohol ketoacidosis, in a 40 y-old chronic alcoholic patient. The detection of trace serum isopropanol prompted a discussion of alcohol ketoacidosis versus toxic isopropanol ingestion or a combination of both, including comparisons with citations in current literature. The automated instruments used to analyze the patient's urine, blood, and serum samples are described. The initial impression was severe metabolic acidosis with an increased anion gap and normal serum glucose and whole blood lactate. Testing for potential toxic ingestions detected only increased serum acetone and trace serum isopropanol. A urinalysis positive for ketones and an increased serum β-hydroxybutyrate concentration clenched the diagnosis of ketoacidosis. Ketoacidosis with an increased anion gap in the absence of hyperglycemia or glycosuria in a chronic alcoholic patient should prompt the evaluation for alcohol ketoacidosis. Trace serum isopropanol may be worrisome for a toxic ingestion, but this finding in severe ketoacidosis may be explained by the reversible action of the enzyme alcohol dehydrogenase. Markedly increased serum isopropanol with a low serum acetone:isopropanol ratio would be more indicative of a toxic isopropanol ingestion.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Isopropanol, 70% in H2O