Merck
  • Synthesis, characterization and potential applications of multifunctional PEO-PPOPEO- magnetic drug delivery system.
Synthesis, characterization and potential applications of multifunctional PEO-PPOPEO- magnetic drug delivery system.
Current medicinal chemistry (2012-12-15)
K Dehvari, K-S Lin
ABSTRACT
Magnetic nanocomposites (MNCs) have highly been acknowledged in the diagnostics and therapeutic applications. Particularly, the multifunctional MNCs have brought a variety of possibilities in targeted drug delivery as well as non-invasive multimodality imaging. A temperature-responsive magnetic drug delivery system has been developed which is made up of superparamagnetic iron oxide nanoparticles (SIONPs) core and Pluronic shell. The magnetic cores composed of congo red conjugated to SIONPs have been proved beneficial as multimodal imaging agents, while superparamagnetic properties facile conducting the nano or micro systems to the vicinity of targeted tissue. Polymer shell formed by stimuli-responsive Pluronic F127 poly(ethylene oxide)-poly(propylene oxide)-poly(ethylene oxide) PEO-PPO-PEO block copolymer serves as the carrier for both hydrophilic and hydrophobic drugs. X-ray diffraction (XRD) and high-resolution transmission electron microscope (HR-TEM) were used to characterize as-synthesized MNCs. Furthermore, vibrating magnetometer experiments showed MNCs having a higher magnetization value than bare magnetic nanoparticles (MNPs) and easy to conduct with an external magnetic field. The hydrodynamic size of MNCs was found to be varying in response to the stimuli temperature. Once the temperature increased, the hydrodynamic radius of MNCs decreased. In addition, the feasibility of the system as a targeted drug delivery system for Alzheimer's diseases (AD) diagnosis and therapy was studied. Searching for reliable targeting molecule, recent approaches for identification of amyloid-β (Aβ) and its derivatives have been evaluated. Consequently, the amyloid-derived diffusible ligands antibodies (anti-ADDLs) have been nominated as potential targeting molecules which can be attached to the MNCs system. The possibility of anti-ADDLs conjugation to DDS has been found promising for the multifunctional drug delivery system for AD diagnosis and therapy. However, further experimental studies are required to assess the performance of the proposed DDS.
Sigma-Aldrich
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Sigma-Aldrich
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Sigma-Aldrich
Iron(III) chloride hexahydrate, ACS reagent, 97%
Sigma-Aldrich
Iron(III) chloride, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Iron(III) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Supelco
Congo Red, analytical standard
Sigma-Aldrich
Iron(II) chloride, 98%
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Congo Red, certified by the BSC, BioXtra
Sigma-Aldrich
Congo Red, Dye content ≥35 %
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)