Merck
  • Doping effects of Co and Cu on superconductivity and magnetism in Fe1+yTe0.6Se0.4 single crystals.
Doping effects of Co and Cu on superconductivity and magnetism in Fe1+yTe0.6Se0.4 single crystals.
Journal of physics. Condensed matter : an Institute of Physics journal (2012-12-15)
Z T Zhang, Z R Yang, L Li, L S Ling, C J Zhang, L Pi, Y H Zhang
ABSTRACT
We report on the investigation of Co and Cu substitution effects on superconductivity and magnetism in Fe(1+y)Te(0.6)Se(0.4) single crystals. The parent Fe(1.01)Te(0.59)Se(0.41) shows a nodeless bulk superconductivity as revealed in heat capacity measurement, which is gradually suppressed by either Co or Cu doping. It is found that the Co or Cu doping mainly serves as scatterers rather than charge carrier doping, which is in agreement with the DFT calculation (2010 Phys. Rev. Lett. 105 157004) reported by Wadati et al. In comparison with Cu doping, Co doping shows a stronger influence on magnetism while a less evident suppression effect on superconductivity. Upon substitution of Co for Fe, a Schottky heat capacity anomaly develops gradually at low temperatures, implying the existence of a paramagnetic moment in the Co-doped samples. In contrast, Cu doping may mainly serve as non-magnetic scatterers, where no Schottky anomaly is observed.
Sigma-Aldrich
Selenium dioxide, 99.999% trace metals basis
Sigma-Aldrich
Selenium dioxide, ReagentPlus®, powder, 99.8% trace metals basis
Sigma-Aldrich
Selenium dioxide, ≥99.9% trace metals basis
Sigma-Aldrich
Selenium dioxide, reagent grade, powder, 98%