Merck
  • Comparison of different advanced oxidation processes for the degradation of two fluoroquinolone antibiotics in aqueous solutions.
Comparison of different advanced oxidation processes for the degradation of two fluoroquinolone antibiotics in aqueous solutions.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2012-12-19)
Maria Bobu, Ayfer Yediler, Ilie Siminiceanu, Feifang Zhang, Sigurd Schulte-Hostede
ABSTRACT
In this study a comparative assessment using various advanced oxidation processes (UV/H(2)O(2), UV/H(2)O(2)/Fe(II), O(3), O(3)/UV, O(3)/UV/H(2)O(2) and O(3)/UV/H(2)O(2)/Fe(II)) was attempted to degrade efficiently two fluoroquinolone drugs ENR [enrofloxacin (1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxo-3-quinolonecarboxylic acid)] and CIP [ciprofloxacin (1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-quinoline-3-carboxylic acid)] in aqueous solutions at a concentrations of 0.15 mM for each drug. The efficiency of the applied oxidation processes (AOPs) has been estimated by the conversion of the original substrate (X(ENR) and X(CIP)) and the reduction of chemical oxygen demand (COD), total organic carbon (TOC). Special emphasis was laid on the effect of varying reaction pH as well as of the applied oxidant doses on the observed reaction kinetics for each advanced oxidation processes. High degradation efficiencies, particularly in terms of rates of TOC and COD abatement, were obtained for photo-Fenton assisted ozonation [O(3)/UV/H(2)O(2)/Fe(II)], compared to other advanced oxidation processes. At pH 3 and 25°C best results for the degradation of both investigated drugs were achieved when 10 mM H(2)O(2), 0.5 mM Fe(II) and an initial dose of 8.5 mg L(-1) ozone were applied. In addition, the evolution of toxicity of the reaction mixtures for different AOPs has been studied by the bioluminescence test (LUMIStox 300).
Product Description
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard