Merck
  • Beta-glucogallin reduces the expression of lipopolysaccharide-induced inflammatory markers by inhibition of aldose reductase in murine macrophages and ocular tissues.
Beta-glucogallin reduces the expression of lipopolysaccharide-induced inflammatory markers by inhibition of aldose reductase in murine macrophages and ocular tissues.
Chemico-biological interactions (2012-12-19)
Kun-Che Chang, Brian Laffin, Jessica Ponder, Anna Enzsöly, János Németh, Daniel V LaBarbera, J Mark Petrash
ABSTRACT
Aldose reductase (AR) catalyzes the reduction of toxic lipid aldehydes to their alcohol products and mediates inflammatory signals triggered by lipopolysaccharide (LPS). Beta-glucogallin (BGG), a recently described AR inhibitor, was purified from extracts of the Indian gooseberry (Emblica officinalis). In this study, we found that BGG showed low cytotoxicity in Raw264.7 murine macrophages and effectively inhibited AR activity as measured by a decrease in sorbitol accumulation. In addition, BGG-mediated inhibition of AR prevented LPS-induced activation of JNK and p38 and lowered ROS levels, which could inhibit LPS-induced apoptosis. Uveitis is a disease of the eye associated with chronic inflammation. In this study, we also demonstrated that treatment with BGG decreased the number of inflammatory cells that infiltrate the ocular media of mice with experimental uveitis. Accordingly, these results suggest BGG is a potential therapy for inflammatory diseases.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.