Merck
  • [Analysis of compositions of the essential oil from Curcuma aromatica by gas chromatography-mass spectrometry].
[Analysis of compositions of the essential oil from Curcuma aromatica by gas chromatography-mass spectrometry].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-12-21)
Ling Chai, Bu-Ming Liu, Xiao Lin, Qi-Xiu Li, Mao-Xiang Lai
ABSTRACT
To analyze the compositions of the essential oil from the rhizome of Curcuma aromatica in Guangxi. The essential oil from the rhizome of Curcuma aromatica was extrated by steam distillation and analysed by GC-MS. 50 chemical constituents accounting for 93.11% of total content were identified. The main components are eucalyptol (53.86%), neocurdione (9.89%), linalool (4.24%), camphor (3.14%), alpha-terpineol (2.94%) and germacrone (2.89%).
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Supelco
(+)-α-Terpineol, analytical standard
Sigma-Aldrich
Eucalyptol, 99%
Supelco
Eucalyptol, analytical standard
Cineole, European Pharmacopoeia (EP) Reference Standard
Supelco
1,8-Cineole, primary reference standard
Sigma-Aldrich
Terpineol, mixture of isomers, 96%, FG
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
Sigma-Aldrich
α-Terpineol, 90%, technical grade