Merck
  • Novel limonene and citral based 2,5-disubstituted-1,3,4-oxadiazoles: a natural product coupled approach to semicarbazones for antiepileptic activity.
Novel limonene and citral based 2,5-disubstituted-1,3,4-oxadiazoles: a natural product coupled approach to semicarbazones for antiepileptic activity.
Bioorganic & medicinal chemistry letters (2012-12-26)
Harish Rajak, Bhupendra Singh Thakur, Avineesh Singh, Kamlesh Raghuvanshi, Anil Kumar Sah, Ravichandran Veerasamy, Prabodh Chander Sharma, Rajesh Singh Pawar, Murli Dhar Kharya
ABSTRACT
Two novel series of N(4)-(5-(2/3/4-substituted-phenyl)-1,3,4-oxadiazol-2-yl)-N(1)-(2-methyl-5-(prop-1-en-2-yl)cyclohex-2-enylidene)semicarbazide and N(4)-(5-(2/3/4-substituted-phenyl)-1,3,4-oxadiazol-2-yl)-N(1)-(3,7-dimethylocta-3,6-dienylidene)-semicarbazide were synthesized to meet structural prerequisite indispensable for anticonvulsant activity. The anticonvulsant activities of the compounds were investigated using maximal electroshock seizure (MES), subcutaneous pentylenetrtrazole (scPTZ) and subcutaneous strychnine (scSTY) models. The rotorod test was conducted to evaluate neurotoxicity. Some of the selected active compounds were subjected to GABA assay to confirm their mode of action. The outcome of the present investigations proved that the four binding sites pharmacophore model is vital for anticonvulsant activity. The efforts were also made to establish structure-activity relationships among test compounds.
Sigma-Aldrich
(R)-(+)-Limonene, 97%
Supelco
(R)-(+)-Limonene, analytical standard
Sigma-Aldrich
(R)-(+)-Limonene, technical, ~90% (sum of enantiomers, GC)
Supelco
(S)-(−)-Limonene, analytical standard
Sigma-Aldrich
(S)-(−)-Limonene, 96%
Sigma-Aldrich
(S)-(−)-Limonene, ≥95%, FG
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Sigma-Aldrich
Citral, 95%
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG