Merck
  • Clinical, microbiological and histological findings in lambs affected by 'salivary abomasum disease'.
Clinical, microbiological and histological findings in lambs affected by 'salivary abomasum disease'.
The Veterinary record (2012-12-27)
G Christodoulopoulos, P R Scott, N Jehl, G Filioussis, S H Smith
ABSTRACT
'Salivary abomasum disease' is a common syndrome in Greece affecting lambs and kids from three to 17 days of age. In this case series, we present clinical and laboratory findings from 37 affected lambs presented alive and subsequently euthanased for welfare reasons and necropsied, and also from 24 other lambs submitted dead that were also necropsied. The clinical signs in the 37 lambs presented alive included lethargy (100 per cent), absence of sucking (83.8 per cent), weakness (37.8 per cent), abdominal distension (40.5 per cent) and increased frequency of urination (24.3 per cent). Diarrhoea was not observed in any affected lambs. At necropsy of these 37 lambs, the abomasum was distended with gas (70.3 per cent), saliva (43.2 per cent) along with mixed milk clots and gastric secretions; while multiple small mucosal and serosal haemorrhages with blood clots ('coffee grains') were recorded (91.9 per cent). Eight of 37 lambs that were examined alive, had elevated blood urea nitrogen concentrations (21.6 per cent). The pH of the abomasal contents ranged from 1.0 to 2.8; Escherichia coli was cultured from six of 37 (16.2 per cent) abomasal fluid samples. A mild to moderate inflammatory cell infiltrate was present in the mucosal lamina propria of 13 of 15 abomasal samples (86.6 per cent). Kidneys were paler than normal in 13 of the total 61 lambs necropsied (21.3 per cent); while acute tubular necrosis was evident on histopathological examination of 11 of 12 examined pale kidneys (91.6 per cent). The low abomasal pH and reported successful treatment with oral sodium bicarbonate suggest that metabolic acidosis may develop during the disease; however, further studies, including blood gas analysis, and determination of D- and L-lactic acid concentrations, are necessary to confirm this hypothesis.
Product Description
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C