Merck
  • Biogenic robust synthesis of silver nanoparticles using Punica granatum peel and its application as a green catalyst for the reduction of an anthropogenic pollutant 4-nitrophenol.
Biogenic robust synthesis of silver nanoparticles using Punica granatum peel and its application as a green catalyst for the reduction of an anthropogenic pollutant 4-nitrophenol.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-01-01)
T Jebakumar Immanuel Edison, M G Sethuraman
ABSTRACT
A robust synthesis of silver nanoparticles (AgNPs) using the peel extract of Punica granatum is reported in this article. The formation of AgNPs was confirmed by the appearance of brownish yellow color and the Surface Plasmon Resonance (SPR) peak at 432 nm. The biogenic AgNPs were found to have the size approximately 30 nm with distorted spherical shape. The high negative zeta potential values of AgNPs revealed their high stability which could be attributed to the capping of AgNPs by the phytoconstituents of the Punica granatum peel. The biogenic AgNPs were also found to function as an effective green catalyst in the reduction of anthropogenic pollutant viz., 4-nitrophenol (4-NP) by solid sodium borohydride, which was evident from the instantaneous color change of bright yellow (400 nm) to colorless (294 nm) solution, after the addition of AgNPs. The catalytic action of biogenic AgNPs in the reduction of 4-NP could be explained on the basis of Langmuir-Hinshelwood model.
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material