Merck
  • Electrophysiological response of chicken's jejunal epithelium to increasing levels of T-2 toxin.
Electrophysiological response of chicken's jejunal epithelium to increasing levels of T-2 toxin.
Mycotoxin research (2013-01-04)
Agha Waqar Yunus, Susan Kröger, Alexander Tichy, Jürgen Zentek, Josef Böhm
ABSTRACT
The present investigations were conducted to test the effects of T-2 toxin on electrophysiological variables of jejunal epithelium of chicken. Jejunal segments of broilers were monitored in Ussing chambers in the presence of T-2 toxin at the levels of 0 (negative control), 0 (methanol/vehicle control), 0.1, 1, 5, and 10 μg/ml of buffer. T-2 toxin did not affect basal values of short circuit current (I(sc)), transmural potential difference, or tissue conductivity in the jejunal epithelium. T-2 toxin also did not statistically affect glucose-induced electrophysiological variables during the first 3 min of glucose induction. Compared to the vehicle control, the ouabain-sensitive I(sc) was negatively affected (P = 0.008) only under 5 μg of T-2 toxin/ml. Increasing levels of T-2 toxin negatively affected the ouabain-sensitive I(sc) in a cubic (P = 0.007) fashion. These data indicate that acute exposure to moderate levels of T-2 toxin may progressively impair the cation gradient across the jejunal epithelium.
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Supelco
T2-Toxin solution, 100 μg/mL in acetonitrile, analytical standard