Merck
BK channels reveal novel phosphate sensitivity in SNr neurons.
PloS one (2013-01-04)
Juan Juan Ji, Lianwan Chen, Xuezhi Duan, Xueqin Song, Wenting Su, Peng Zhang, Li Li, Shuyun Bai, Yingchun Sun, Nobuya Inagaki
ABSTRACT
Whether large conductance Ca(2+)-activated potassium (BK) channels are present in the substantia nigra pars reticulata (SNr) is a matter of debate. Using the patch-clamp technique, we examined the functional expression of BK channels in neurons of the SNr and showed that the channels were activated or inhibited by internal high-energy phosphates (IHEPs) at positive and negative membrane potentials, respectively. SNr neurons showed membrane potential hyperpolarization under glucose-deprivation conditions which was attenuated by paxilline, a specific BK channel blocker. In addition, Fluo-3 fluorescence recording detected an increase in the level of internal free calcium ([Ca(2+)](i)) during ischemic hyperpolarization. These results confirm that BK channels are present in SNr neurons and indicate that their unique IHEP sensitivity is requisite in neuronal ischemic responses. Bearing in mind that the K(ATP) channel blocker tolbutamide also attenuated the hyperpolarization, we suggest that BK channels may play a protective role in the basal ganglia by modulating the excitability of SNr neurons along with K(ATP) channels under ischemic stresses.
Sigma-Aldrich
Paxilline, powder, ≥98% (HPLC)