Merck
  • Smart mesoporous SiO2 nanoparticles for the DNAzyme-induced multiplexed release of substrates.
Smart mesoporous SiO2 nanoparticles for the DNAzyme-induced multiplexed release of substrates.
Journal of the American Chemical Society (2013-01-10)
Zhanxia Zhang, Dora Balogh, Fuan Wang, Itamar Willner
ABSTRACT
The fluorescent dyes methylene blue, MB(+), and thionine, Th(+), can be trapped in the pores of mesoporous silica, MP-SiO(2), by means of functional nanostructures consisting of the Mg(2+)- or Zn(2+)-dependent DNAzyme sequences. In the presence of Mg(2+) or Zn(2+) ions the respective DNAzymes are activated, leading to the specific cleavage of the respective caps, and the selective release of MB(+) or Th(+). The enlargement of the conserved loop domains of the Mg(2+)- or Zn(2+)-dependent DNAzyme sequences with foreign nucleotides prohibits the formation of active DNAzymes and eliminates the release of the respective dyes. This is due to the flexibility of the loops that lacks affinity for the association of the ions. The insertion of aptamer sequences (e.g., the adenosine-5'-triphosphate (ATP) aptamer) or ion-binding sequences (e.g., T-rich Hg(2+) ion-binding domains) as foreign components to the loop regions allows the formation of active Mg(2+)- or Zn(2+)-dependent DNAzyme structures through the cooperative formation of aptamer-ATP complexes or T-Hg(2+)-T bridges. These aptamer-substrate complexes or T-Hg(2+)-T bridges allosterically stabilize and activate the DNAzymes, thus allowing the selective release of the fluorescent substrates MB(+) or Th(+). The metal ion-driven DNAzyme release of substrates from the pores of MP-SiO(2), and particularly the allosteric activation of the DNAzymes through cooperative aptamer-substrate complexes or metal-ion bridges, has important future nanomedical implications for targeted release of drugs. This is demonstrated with the triggered release of the anticancer drug, doxorubicin, by the Mg(2+)-DNAzyme-locked pores or by the aptamer-ATP complex-triggered activation of the Mg(2+)-dependent DNAzyme.
Sigma-Aldrich
Thionin acetate salt, certified by the BSC, Dye content ≥85 %