Merck
  • Lactobacillus johnsonii inhibits indoleamine 2,3-dioxygenase and alters tryptophan metabolite levels in BioBreeding rats.
Lactobacillus johnsonii inhibits indoleamine 2,3-dioxygenase and alters tryptophan metabolite levels in BioBreeding rats.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-01-11)
Ricardo Valladares, Lora Bojilova, Anastasia H Potts, Evan Cameron, Christopher Gardner, Graciela Lorca, Claudio F Gonzalez
ABSTRACT
In our previous work, we found that feeding Lactobacillus johnsonii to BioBreeding diabetes-prone (BBDP) rats decreased the incidence of diabetes development. The aim of this study was to investigate host pathways affected by L. johnsonii, with specific focus on the rate-limiting enzyme of tryptophan catabolism, indoleamine 2,3-dioxygenase (IDO). Suspensions of L. johnsonii or an equal volume of vehicle were orally administered to BBDP rats. Tissue IDO was investigated using quantitative RT-PCR and Western blot, whereas tryptophan, kynurenine, and 5-hydroxytryptamine (5-HT) concentrations were quantified by HPLC and ELISA. IDO activity was also investigated using L. johnsonii culture cell-free supernatant (CFS) with affinity-purified IDO and HT-29 intestinal epithelial cells. L. johnsonii feeding resulted in a 17% reduction in serum kynurenine compared with that in vehicle-fed controls, correlating with a 1.4-fold elevation in 5-HT levels. H₂O₂ produced by L. johnsonii abolished IDO activity in vitro, and L. johnsonii feeding resulted in a 3.9-fold increase in ileum lumen H₂O₂. L. johnsonii CFS significantly reduced IDO activity in HT-29 intestinal epithelial cells (47% reduction) compared with that in vehicle-treated controls, an effect abolished by catalase treatment. These data support the role of H₂O₂ in commensal bacteria-host interactions and highlight the influence of commensal bacteria-derived H₂O₂ on host physiology.
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
DL-Kynurenine, ≥95.0% (NT)
Sigma-Aldrich
Indole, ≥99%, FG