Merck
  • A simple metal-promoted three-step access to n/5/m angular carbocyclic systems.
A simple metal-promoted three-step access to n/5/m angular carbocyclic systems.
Organic letters (2013-01-12)
José Barluenga, Ana Álvarez-Fernández, Tatiana Suárez-Rodríguez, Ángel L Suárez-Sobrino, Miguel Tomás
ABSTRACT
A simple three-step access to n/5/m angular tricyclic skeleta from terminal cycloalkenylacetylenes, Cr(CO)(6), and propargylmalonate esters is described. The process involves a cyclopentannulation of propargylmalonates and enynyl Fischer carbenes, followed by a metal-promoted Cope-type fragmentation of the propargyl unit. The resulting 4-allenyl-carbonyl derivatives can undergo either a 5-exo annulation via a tin enolate or an unprecedented gold catalyzed 6-endo cyclization.
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis