Merck
A novel biphasic oral contrast solution for enterographic studies.
Journal of computer assisted tomography (2013-01-17)
Oktay Algin, Sehnaz Evrimler, Evrim Ozmen, Melike Ruşen Metin, Gokhan Ocakoglu, Osman Ersoy, Mustafa Karaoglanoglu, Halil Arslan
ABSTRACT
To compare the efficacy and patients' tolerance of a new mixed biphasic oral contrast solution with routine biphasic oral contrast agent in magnetic resonance (MR) enterography (MRE). Thirty-seven patients (group 1) had MRE with the new mixture, whereas 14 patients (group 2) had MRE with biphasic oral contrast agent (lactulose). Magnetic resonance enterography images were evaluated by 2 experienced radiologists. Each intestinal segment was evaluated for luminal distension (LD), distinction from the surrounding tissue (wall conspicuity), and the confidence of radiologist for evaluation of the specified segment (radiological evaluation confidence). Comparisons between the 2 groups were performed using the Mann-Whitney U test. Interrater and intrarater agreement values were obtained. In addition, patients' acceptability and tolerance were assessed. The new mixture was more effective than the oral contrast agent used in group 2 for LD, wall conspicuity, and radiological confidence. The values of interrater and intrarater agreement in scoring LD, wall conspicuity, and radiological confidence were generally moderate. Our new mixture allowed good-quality enterographic images, and this solution was well tolerated by patients. In addition, this mixture is useful for evaluation of small bowels and colonic segments. We suggest the use of it for enterographic examinations.
Product Description
Lactulose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lactulose, ≥98.0% (HPLC)
Sigma-Aldrich
Barium sulfate, meets USP testing specifications
Sigma-Aldrich
Barium sulfate, ReagentPlus®, 99%
Sigma-Aldrich
Barium sulfate, 99.99% trace metals basis
Sigma-Aldrich
Lactulose, ≥95% (HPLC)
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP