Merck
  • Clathrin-mediated hemoglobin endocytosis is essential for survival of Leishmania.
Clathrin-mediated hemoglobin endocytosis is essential for survival of Leishmania.
Biochimica et biophysica acta (2013-01-19)
Shruti Agarwal, Ruchir Rastogi, Deepika Gupta, Nitin Patel, Manoj Raje, Amitabha Mukhopadhyay
ABSTRACT
Leishmania is auxotroph for heme. Previously, we have shown that Leishmania acquire heme from the degradation of endocytosed hemoglobin via a specific receptor located in the flagellar pocket. Here, we report the cloning and expression of clathrin heavy chain from Leishmania (Ld-CHC) and provide evidences that Ld-CHC is localized in flagellar pocket and regulates Hb-endocytosis in Leishmania. Kinetic analysis of Hb trafficking in GFP-Ld-CHC overexpressed Leishmania reveals that Hb is internalized through Ld-CHC coated region and remains associated with Ld-CHC containing vesicles at early time points of internalization and subsequently starts dissociating from Hb-containing vesicles at later time points indicating that clathrin-coating and uncoating regulate Hb trafficking in Leishmania. Interestingly, overexpression of dominant negative mutant of clathrin heavy chain of Leishmania (GFP-Ld-CHC-Hub) blocks the Hb internalization and causes severe growth defect in parasite. Moreover, we have shown that chlorpromazine, a pharmacological agent, blocks Hb internalization in Leishmania by depolymerizing Ld-CHC and thereby inhibits the growth of the parasites. Taken together, our results have shown that Hb endocytosis in Leishmania is a clathrin dependent process and is essential for the survival of the parasites.
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Supelco
Chlorpromazine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard