Merck
  • Curcumin protects against iron induced neurotoxicity in primary cortical neurons by attenuating necroptosis.
Curcumin protects against iron induced neurotoxicity in primary cortical neurons by attenuating necroptosis.
Neuroscience letters (2013-01-19)
Min-Chao Dai, Zhi-Hong Zhong, Yu-Hao Sun, Qing-Fang Sun, Yong-Ting Wang, Guo-Yuan Yang, Liu-Guan Bian
ABSTRACT
Necroptosis was reported as one backup way of programmed cell death when apoptosis was blocked, and the receptor interacting protein 1 was considered as the key necroptosis regulator protein. Here, we report the neuroprotective effects of curcumin which attenuates necroptosis. Primary cortical neurons were cultured and were injured by ferrous chloride, z.vad.fmk was applied to block apoptosis, curcumin was administrated to protect neurons, necrostatin-1 was applied to inhibit necroptosis if needed. Cell viability was measured by detecting lactate dehydrogenase activity in lysates of surviving cells, and assessed by cell counting kit-8. The expression of receptor interacting protein 1 was detected by immunoblot and immunofluorescence. Results showed that necroptosis mainly occurred in the concentrations of ferrous chloride ranging from 100 to 200μM, curcumin attenuated necroptosis in a dose-dependent manner. Furthermore, curcumin decreased expression of receptor interacting protein 1 in a dose- and time-dependent manner. Taken together, these findings suggest that curcumin protects against iron induced neurotoxicity in primary cortical neurons by attenuating necroptosis.
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%