Merck
  • Purification and characterization of an extracellular keratinolytic protease from a new isolate of Aspergillus parasiticus.
Purification and characterization of an extracellular keratinolytic protease from a new isolate of Aspergillus parasiticus.
Protein expression and purification (2013-01-23)
T S Anitha, P Palanivelu
ABSTRACT
Keratinolytic proteases find extensive applications both in environmental biotechnology and pharmaceutical industries. An extracellular keratinolytic protease was purified and characterized from the fungus, Aspergillus parasiticus, isolated from poultry soil. The enzyme was purified to homogeneity by acetone and ammonium sulfate precipitations followed by CM-Sepharose column chromatography. The molecular mass of the enzyme was 36kDa as judged by SDS-PAGE. The purified keratinase had a pH optimum of 7.0 and temperature optimum of 50(o)C. The enzyme hydrolyzed the substrate azocasein and the Km and Vmax of the purified keratinase were found to be 1.04mg/ml and 3463.34Units/min/mg protein, respectively. The enzyme showed increased activity in the presence of reducing agents. The enzyme was found to be glycosylated. According to the inhibition profiles obtained with the various protease inhibitors, it was confirmed that the purified keratinase belongs to the serine protease type. The purified enzyme activity was enhanced by calcium, magnesium and manganese ions and partially inhibited by cadmium, copper and zinc ions. The purified enzyme showed increased activity with nonionic detergents and urea.
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%