Merck
  • V101L of human formyl peptide receptor 1 (FPR1) increases receptor affinity and augments the antagonism mediated by cyclosporins.
V101L of human formyl peptide receptor 1 (FPR1) increases receptor affinity and augments the antagonism mediated by cyclosporins.
The Biochemical journal (2013-02-05)
Caihong Zhou, Yan Zhou, Jia Wang, Yang Feng, Haonan Wang, Jinglun Xue, Yani Chen, Richard D Ye, Ming-Wei Wang
ABSTRACT
Genetic variation plays a major role in drug response variability. CsA (cyclosporin A), a widely used immunosuppressive agent, is a specific antagonist for FPR1 (formyl peptide receptor 1), which is an important G-protein-coupled chemoattractant receptor in the innate immune system. In order to study the variable responses of cyclosporins to different FPR1 mutants, we investigated the distribution of human FPR1 haplotypes among 209 healthy Han Chinese subjects. The haplotype pattern in Han Chinese were characterized on the basis of five SNPs (single nucleotide polymorphisms), including rs5030878 (p.T11I), rs2070745 (p.V101L), rs5030880 (p.R190W), rs1042229 (p.N192K) and rs867228 (p.A346E). Receptor binding affinity of cyclosporins to FPR1 haplotypes was assessed using N-formyl-Nle-Leu-Phe-Nle-Tyr-Lys-FITC in CHO-G(α16) cells stably transfected with cDNAs encoding the top 12 FPR1 haplotypes in the Han Chinese. Variants of FPR1 carrying a single amino acid substitution of leucine for valine at position 101 (p.Leu(101)) displayed significantly higher pK(i) values for CsA and CsH (cyclosporin H), indicative of an improved receptor affinity. The polymorphism of FPR1 p.Leu(101) also enhanced the inhibitory effects of cyclosporins on fMLF (N-formyl-methionyl-leucyl-phenylalanine)-induced activities, including calcium mobilization, cell chemotaxis and MAPK (mitogen-activated protein kinase) phosphorylation. These results point to a possible complication for clinical use of CsA in patients carrying the p.Leu(101) allele of FPR1.
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, ≥97% (HPLC)