Merck
  • A split-faced, observer-blinded comparison study of topical adapalene/benzoyl peroxide and adapalene in the treatment of Asian acne patients.
A split-faced, observer-blinded comparison study of topical adapalene/benzoyl peroxide and adapalene in the treatment of Asian acne patients.
Journal of drugs in dermatology : JDD (2013-02-05)
Won-Jeong Kim, Jung-Min Park, Hyun-Chang Ko, Byung-Soo Kim, Moon-Bum Kim, Margaret Song
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water