Merck
  • Design, modification and 3D QSAR studies of novel naphthalin-containing pyrazoline derivatives with/without thiourea skeleton as anticancer agents.
Design, modification and 3D QSAR studies of novel naphthalin-containing pyrazoline derivatives with/without thiourea skeleton as anticancer agents.
Bioorganic & medicinal chemistry (2013-02-09)
Wen Yang, Yang Hu, Yu-Shun Yang, Fei Zhang, Yan-Bin Zhang, Xiao-Liang Wang, Jian-Feng Tang, Wei-Qing Zhong, Hai-Liang Zhu
ABSTRACT
Two series of novel naphthalin-containing pyrazoline derivatives C1-C14 and D1-D14 have been synthesized and evaluated for their EGFR/HER-2 inhibitory and anti-proliferation activities. Compound D14 displayed the most potent activity against EGFR and A549 cell line (IC(50)=0.05 μM and GI(50)=0.11 μM), being comparable with the positive control Erlotinib (IC(50)=0.03 μM and GI(50)=0.03 μM) and more potent than our previous compounds C0-A (IC(50)=5.31 μM and GI(50)=33.47 μM) and C0-B (IC(50)=0.09 μM and GI(50)=0.34 μM). Meanwhile, compound C14 displayed the most potent activity against HER-2 and MCF-7 cell line (IC(50)=0.88 μM and GI(50)=0.35 μM), being a little less potent than Erlotinib (IC(50)=0.16 μM and GI(50)=0.08 μM) but far more potent than C0-A (IC(50)=6.58 μM and GI(50)=27.62 μM) and C0-B (IC(50)=2.77 μM and GI(50)=3.79 μM). The docking simulation was performed to analyze the probable binding models and the QSAR models were built for reasonable design of EGFR/HER-2 inhibitors at present and in future. The structural modification of introducing naphthalin moiety reinforced the combination of our compounds and the receptor, resulting in progress of bioactivity. Moreover, the replacement of thiourea skeleton by using benzene ring resulted in the slight diversity of the two series towards specific targets.
Product Description
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Melting point standard 79-81°C, analytical standard