Merck
  • Comparative evaluation of taurine and thiotaurine as protectants against diabetes-induced nephropathy in a rat model.
Comparative evaluation of taurine and thiotaurine as protectants against diabetes-induced nephropathy in a rat model.
Advances in experimental medicine and biology (2013-02-09)
Kashyap G Pandya, Roshil Budhram, George Clark, Cesar A Lau-Cam
ABSTRACT
Taking into account the proven effectiveness of antioxidants in preventing experimentally induced diabetes in laboratory animals, this study was carried out with the specific purpose of comparing the effectiveness of two known antioxidants, the β-aminosulfonate taurine (TAU) and β-aminothiosulfonate thiotaurine (TTAU), in preventing biochemical, functional and histological alterations indicative of -diabetic nephropathy. In the study, streptozotocin (60 mg/kg, orally) was used to induce type 2 diabetes mellitus in Sprague-Dawley rats. Starting on day 15 and continuing up to day 56, the rats received a daily single 2.4 mmol/kg oral dose of a sulfur-containing compound (TAU or TTAU) or 4 U/kg subcutaneous dose of isophane insulin (INS). Rats not receiving any treatment served as controls. After obtaining a 24 h urine sample, the animals were sacrificed by decapitation on day 57, and their blood and kidneys immediately collected. Diabetic rats exhibited marked hyperglycemia, hypoinsulinemia, hypoproteinemia, hyponatremia, hyperkalemia, azotemia, hypercreatinemia, increased plasma TGF β(1), lipid peroxidation, plasma and kidney nitrite, and urine output; decreased glutathione redox status in plasma and kidney, decreased urine Na(+) and K(+), proteinuria and hypocreatinuria. Without exceptions, all the treatment compounds were found to markedly and variously attenuate these changes. Confirmation of protection by INS, TAU and TTAU was provided by the results of histological examination of kidney sections and which showed a more normal appearance than sections from diabetic animals. In most instances protection by TTAU was about equal to that by INS but greater than that by TAU.
Product Description
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, ≥98%, FG
SAFC
Taurine
Sigma-Aldrich
L-Glutathione oxidized, BioXtra, ≥98%
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized, ≥98%, lyophilized powder
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%