Merck
  • A Phase I study of arsenic trioxide (Trisenox), ascorbic acid, and bortezomib (Velcade) combination therapy in patients with relapsed/refractory multiple myeloma.
A Phase I study of arsenic trioxide (Trisenox), ascorbic acid, and bortezomib (Velcade) combination therapy in patients with relapsed/refractory multiple myeloma.
Cancer investigation (2013-02-15)
Lauren A Held, David Rizzieri, Gwynn D Long, Jon P Gockerman, Louis F Diehl, Carlos M de Castro, Joseph O Moore, Mitchell E Horwitz, Nelson J Chao, Cristina Gasparetto
ABSTRACT
This Phase I study assessed the feasibility of concomitant arsenic trioxide (ATO), ascorbic acid (AA), and bortezomib (Velcade™) (AAV) for patients with relapsed/refractory multiple myeloma. ATO (0.25 mg/kg) and AA (1 g) were given with an escalating dose of bortezomib (1 mg/m(2) or 1.3 mg/m(2) IV bolus on days 1 and 8 of a 21-day cycle). Ten patients (median age 62 years), with a median of 3 prior regimens, were enrolled. Four (40%) patients achieved clinical benefit, with one patient achieving a durable partial response. No formal DLTs were encountered. AAV combination was feasible and demonstrated some benefits in this heavily pretreated population.
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis
Supelco
Arsenic(III) oxide, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Arsenic(III) oxide, ACS reagent (primary standard)
Sigma-Aldrich
Arsenic(III) oxide, ReagentPlus®, ≥99.0%