Merck
  • RNA interference against discoidin domain receptor 2 ameliorates alcoholic liver disease in rats.
RNA interference against discoidin domain receptor 2 ameliorates alcoholic liver disease in rats.
PloS one (2013-02-15)
Zheng Luo, Huimin Liu, Xiaomeng Sun, Rong Guo, Ruibing Cui, Xiangxing Ma, Ming Yan
ABSTRACT
Discoidin domain receptor 2 (DDR2) is involved in fibrotic disease. However, the exact pathogenic implications of the receptor in early alcoholic liver disease are still controversial. We constructed plasmid vectors encoding short-hairpin RNA against DDR2 to investigate its role in alcoholic liver disease in an immortalized rat hepatic stellate cell line, HSC-T6, and in rats by MTT, RT-PCR and western blot analyses; immunohistochemistry and electron microscopy. Alcohol-induced upregulation of DDR2 was associated with the expression of matrix metalloproteinase 2, the transforming growth factor β1 signaling pathway and tissue inhibitor of metalloproteinase 1; collagen deposition; and extracellular matrix remodeling. Inhibition of DDR2 decreased HSC-T6 cell proliferation and liver injury in rats with 10-week-induced alcoholic liver disease. DDR2 may have an important role in the pathogenesis of early-stage alcoholic liver disease. Silencing DDR2 may be effective in preventing early-stage alcoholic liver disease.
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF