Merck
  • Effects of amylosucrase treatment on molecular structure and digestion resistance of pre-gelatinised rice and barley starches.
Effects of amylosucrase treatment on molecular structure and digestion resistance of pre-gelatinised rice and barley starches.
Food chemistry (2013-02-16)
Bum-Su Kim, Hyun-Seok Kim, Jung-Sun Hong, Kerry C Huber, Jae-Hoon Shim, Sang-Ho Yoo
ABSTRACT
Structural modification of rice and barley starches with Neisseria polysaccharea amylosucrase (NpAS) was conducted, and relationship between structural characteristics and resistant starch (RS) contents of NpAS-treated starches was investigated. Pre-gelatinised rice and barley starches were treated with NpAS. NpAS-treated starches were characterised with respect to morphology, X-ray diffraction pattern, amylopectin branch-chain distribution, and RS content, and their structural characteristics were correlated to RS contents. Regardless of amylose contents of native starches, NpAS-treated (relative to native) starches possessed lower and higher proportions of shorter (DP 6-12) and intermediate (DP 13-36) amylopectin (AP) branch-chains, respectively. RS contents were higher for NpAS-treated starches relative to native starches, and maximum RS contents were obtained for NpAS-treated starches of waxy rice and barley genotypes. Amylose contents were not associated with RS contents of NpAS-treated starches. However, shorter and intermediate AP branch-chain portions were negatively and positively correlated to RS contents of NpAS-treated starches, respectively.
Sigma-Aldrich
Amylose from potato, used as amylase substrate