Merck
  • Vibrational (FT-IR and FT-Raman) spectra and quantum chemical studies on the molecular orbital calculations, chemical reactivity and thermodynamic parameters of 2-chloro-5-(trifluoromethyl) aniline.
Vibrational (FT-IR and FT-Raman) spectra and quantum chemical studies on the molecular orbital calculations, chemical reactivity and thermodynamic parameters of 2-chloro-5-(trifluoromethyl) aniline.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-19)
T Karthick, V Balachandran, S Perumal, A Nataraj
ABSTRACT
In this work, the vibrational characteristics of 2-chloro-5-(trifluoromethyl) aniline have been investigated and both the experimental and theoretical vibrational data indicate the presence of various functional groups within the title molecule. The influence of chlorine substituent on the vibrational wavenumbers of a molecule in comparison with aniline and trifluoromethyl aniline has been discussed in detail. The density functional theoretical (DFT) computations were performed at the B3LYP/6-31++G(3df,3pd)/6-31G(3df,3pd) levels to derive the optimized geometry, vibrational wavenumbers with IR and Raman intensities. Furthermore, the molecular orbital calculations such as; natural bond orbitals (NBOs) and HOMO-LUMO energy gap and mapped molecular electrostatic potential (MEP) surfaces were also performed with the same level of DFT. The temperature dependence thermodynamic parameters of a molecule were illustrated on the basis of their correlation graphs. The detailed interpretation of the vibrational spectra has been carried out with the aid of potential energy distribution (PED) results obtained from MOLVIB program. The delocalization of electron density in various constituents of the molecule has been discussed with the aid of NBO and HOMO-LUMO energy gap analysis.
Sigma-Aldrich
2-Chloro-4-(trifluoromethyl)aniline, 97%
Pricing and availability is not currently available.