Merck
  • Evaluation of phytotoxicity and genotoxicity of nitrobenzene with a battery of Vicia faba assay system.
Evaluation of phytotoxicity and genotoxicity of nitrobenzene with a battery of Vicia faba assay system.
Environmental toxicology and chemistry (2013-02-21)
Jun Ma, Donglin Guo, Wenyue Su, Dan Wang, Changhong Guo
ABSTRACT
Nitrobenzene (NB) is an important organic compound intermediate that is used widely in industry. In the present study, to evaluate the phytotoxicity and genotoxicity of NB on plants, Vicia faba was exposed to increasing concentrations of NB (5 mg L(-1) , 10 mg L(-1) , 25 mg L(-1) , 50 mg L(-1) , and 100 mg L(-1) ). The data revealed that germination rate and radicle length of V. faba seedlings were promoted by low NB concentrations and short exposure periods, whereas these parameters were inhibited at greater NB concentrations and longer exposures. When assessed by mitotic index, micronucleus, and chromosomal aberration assays, NB showed dose-dependent genotoxicity at 0 mg L(-1) to 50 mg L(-1).
Product Description
Supelco
Nitrobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Nitrobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Nitrobenzene, ACS reagent, ≥99.0%