Merck
  • Modulation of the intestinal environment, innate immune response, and barrier function by dietary threonine and purified fiber during a coccidiosis challenge in broiler chicks.
Modulation of the intestinal environment, innate immune response, and barrier function by dietary threonine and purified fiber during a coccidiosis challenge in broiler chicks.
Poultry science (2013-02-26)
E L Wils-Plotz, M C Jenkins, R N Dilger
ABSTRACT
Coccidiosis is a major contributor to economic losses in the poultry industry due to its detrimental effects on growth performance and nutrient utilization. We hypothesized that the combined effects of supplemental dietary Thr and purified fiber may modulate the intestinal environment and positively affect intestinal immune responses and barrier function in broiler chicks infected with Eimeria maxima. A Thr-deficient basal diet (3.1 g of Thr/kg of diet) was supplemented with 70 g/kg of silica sand (control) or high-methoxy pectin and 1 of 2 concentrations of Thr (1.8 or 5.3 g/kg of diet; 4 diets total), and fed to chicks from hatch to d 16 posthatch. On d 10 posthatch, chicks received 0.5 mL of distilled water or an acute dose of Eimeria maxima (1.5 × 10(3) sporulated oocytes) with 6 replicate pens of 6 chicks per each of 8 treatment combinations (4 diets and 2 inoculation states). Body weight gain, feed intake, and G:F increased (P < 0.01) with addition of 5.3 g of Thr/kg of diet. Eimeria maxima schizonts were present only in intestinal tissue sampled from infected birds (P < 0.01). Weights of cecal digesta were highest (P < 0.01) in pectin-fed birds, and ceca with the heaviest weights also had the highest concentrations of total short-chain fatty acids. Expression of interleukin-12 in ileal mucosa was highest (P < 0.01) in infected birds receiving the control diet with 5.3 g of supplemental Thr/kg. In cecal tonsils, interferon-γ expression was highest in infected birds receiving the control diet (fiber × infection, P < 0.05); interferon-γ expression was lowest in infected birds fed the high Thr diet (Thr × infection, P < 0.05). There were no differences due to infection or Thr supplementation for cytokine expression in birds fed pectin-containing treatments. Overall, we conclude that although pectin has some protective function against coccidiosis, Thr supplementation had the greatest effect on intestinal immune response and maintenance of near normal growth in young broiler chicks infected with E. maxima.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material