Merck
  • Surface functionality as a means to impact polymer nanoparticle size and structure.
Surface functionality as a means to impact polymer nanoparticle size and structure.
Langmuir : the ACS journal of surfaces and colloids (2013-02-27)
Julia Schneider, Andrew P Jallouk, Daniela Vasquez, Ralf Thomann, Aurélien Forget, Christopher J Pino, V Prasad Shastri
ABSTRACT
When polymeric nanoparticles (NPs) are formed by nanoprecipitation, which is a nucleation-growth process, the control over size requires changing the polymer concentration or solvent composition. Here, we demonstrate that the NP size can be controlled independent of polymer variables by introducing a polyelectrolyte (PE) in the aqueous phase. PEs that exhibit hydrogen bonding (H-bonding) yield a reduction in NP size, whereas PEs that do not possess this characteristic promote the formation of larger NPs. The observed effect can be attributed to the formation of a diffusional barrier around the NP in the form of a dense shell. This principle of controlling NP size is not limited to polymers and can also be employed in the production of lipid NPs.
Sigma-Aldrich
Polycaprolactone, average Mn 80,000
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
Sigma-Aldrich
Polycaprolactone, average Mw ~14,000, average Mn ~10,000 by GPC
Sigma-Aldrich
Polycaprolactone, average Mn 45,000
Sigma-Aldrich
Poly(4-styrenesulfonic acid) solution, Mw ~75,000, 18 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~1,000,000, powder
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
Supelco
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~70,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~4,000,000
Sigma-Aldrich
Poly(acrylic acid)
Sigma-Aldrich
Poly(acrylic acid), average Mv ~1,250,000
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~70,000, 30 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~1,000,000, 25 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Sigma-Aldrich
Poly(acrylic acid), average Mv ~3,000,000
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~200,000, 30 wt. % in H2O
Supelco
Poly(acrylic acid sodium salt), analytical standard, suitable for gel permeation chromatography (GPC), 16,000
Supelco
Poly(acrylic acid sodium salt), analytical standard, suitable for gel permeation chromatography (GPC), 1,250