Merck
  • Long-acting human serum albumin-thioredoxin fusion protein suppresses bleomycin-induced pulmonary fibrosis progression.
Long-acting human serum albumin-thioredoxin fusion protein suppresses bleomycin-induced pulmonary fibrosis progression.
The Journal of pharmacology and experimental therapeutics (2013-02-28)
Ryota Tanaka, Hiroshi Watanabe, Azusa Kodama, Victor Tuan Giam Chuang, Yu Ishima, Keisuke Hamasaki, Ken-ichiro Tanaka, Tohru Mizushima, Masaki Otagiri, Toru Maruyama
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is thought to involve inflammatory cells and reactive oxygen species (ROS), such as superoxide anion radical (O2(·-)). There is currently no effective treatment of IPF. We previously developed a human serum albumin (HSA)-thioredoxin 1 (Trx) fusion protein (HSA-Trx) designed to overcome the unfavorable pharmacokinetic and short pharmacological properties of Trx, an antioxidative and anti-inflammatory protein. In this study, we examined the therapeutic effect of HSA-Trx on an IPF animal model of bleomycin (BLM)-induced pulmonary fibrosis. A pharmacokinetic study of HSA-Trx or Trx in BLM mice showed that the plasma retention and lung distribution of Trxc was markedly improved by fusion with HSA. A weekly intravenous administration of HSA-Trx, but not Trx, ameliorated BLM-induced fibrosis, as evidenced by a histopathological analysis and pulmonary hydroxyproline levels. HSA-Trx suppressed active-transforming growth factor (TGF)-β levels in the lung and inhibited the increase of inflammatory cells in bronchoalveolar lavage fluid, pulmonary inflammatory cytokines, and oxidative stress markers. An in vitro EPR experiment using phosphate-buffered saline-stimulated neutrophils confirmed the O2(·-) scavenging ability of HSA-Trx. Furthermore, post-treatment of HSA-Trx had a suppressive effect against BLM-induced fibrosis. These results suggest that HSA-Trx has potential as a novel therapeutic agent for IPF, because of its long-acting antioxidative and anti-inflammatory modulation effects.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Thioredoxin from Escherichia coli, recombinant, expressed in E. coli, essentially salt-free, lyophilized powder, ≥3 units/mg protein
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, suitable for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioReagent, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Supelco
trans-4-Hydroxy-L-proline, analytical standard