Merck
Effect of artificial aging on the bond durability of fissure sealants.
The journal of adhesive dentistry (2013-03-28)
Xiaofei Yun, Wei Li, Chen Ling, Alex Fok
ABSTRACT
To evaluate the effect of artificial aging on the bond durability of fissure sealants in vitro. Twenty bovine incisors received 4 different sealant treatments and were divided into four groups: 1. Ultraseal XT plus (UX); 2. Enamel Loc (EL); 3. 35% phosphoric acid plus Enamel Loc (PEL); 4. Adper Prompt L-Pop plus Clinpro (PPC). Beam-shaped specimens were prepared and randomly divided into three subgroups. One subgroup underwent the microtensile bond strength (µTBS) test after 24-h storage in 37°C water. The other two subgroups were also subjected to the microtensile bond strength test after 5000 and 10,000 thermal cycles, respectively. Another twelve intact human third molars were sealed using 1 of 3 methods and were divided into 3 groups of 4 each: 1. Ultraseal XT plus; 2. Adper Prompt L-Pop plus Clinpro; and 3. Enamel Loc. Two specimens from each group were immersed in a 50% silver nitrate solution for 24 h, followed by exposure to fluorescent light for 8 h, before being scanned in a micro-CT (microcomputer tomography) machine. The other two were handled in the same way after undergoing 10,000 thermal cycles. The CT images obtained were evaluated. All samples from the EL group were broken during preparation, so no µTBS results were available. After 5000 thermal cycles, the bond strengths of the three other groups (UX, PEL, PPC) decreased significantly (p < 0.05). Longer thermocycling (10,000 cycles) resulted in more decreases in µTBS for group PEL and PPC, while the strength of the UX group remained relatively unchanged. After thermocycling, considerable silver penetration could be seen at the sealant/enamel interface of the EL group in micro-CT images. The etch-and-rinse procedure for sealant application promotes higher bond strength under artificial aging. Micro-CT, a nondestructive analytical tool, may be used to evaluate the sealant/enamel interface effectively.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 80-120 ppm MEHQ as inhibitor, 95%
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)