Merck
  • Carbonic anhydrase IX (CAIX) is not an independent predictor of outcome in patients with clear cell renal cell carcinoma (ccRCC) after long-term follow-up.
Carbonic anhydrase IX (CAIX) is not an independent predictor of outcome in patients with clear cell renal cell carcinoma (ccRCC) after long-term follow-up.
BJU international (2013-04-05)
Ben Y Zhang, R Houston Thompson, Christine M Lohse, Roxana S Dronca, John C Cheville, Eugene D Kwon, Bradley C Leibovich
ABSTRACT
To evaluate carbonic anhydrase IX (CAIX) expression as an independent prognostic marker for clear cell renal cell carcinoma (ccRCC). With recent smaller studies showing conflicting results, we aimed to update our initial analysis in 2007 with an additional 5-year follow-up. We provided long-term follow-up of the same cohort used in our 2007 study (730 patients with unilateral, sporadic ccRCC treated surgically between 1990 and 1999). Associations of CAIX expression with RCC death and distant metastases were evaluated using Cox proportional hazards regression models. CAIX was expressed in 708 (97.0%) of the specimens; 163 tumours (22.3%) had low (≤85%) expression and 567 (77.7%) high (>85%) expression. There were 483 deaths and 265 RCC-specific deaths. The median follow-up for the 247 patients still under observation was 13.8 years. Univariately, low CAIX expression was associated with an increased risk of RCC death vs high expression (hazard ratio 1.62; P < 0.001). Low CAIX expression was not statistically significantly associated with RCC death or distant metastases after adjusting for nuclear grade or coagulative tumour necrosis. After additional long-term follow-up of our large cohort, our results continue to suggest that CAIX is not an independent prognostic marker for ccRCC.
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6