Merck
  • Comprehensive review and compilation of treatment for azo dyes using microbial fuel cells.
Comprehensive review and compilation of treatment for azo dyes using microbial fuel cells.
Water environment research : a research publication of the Water Environment Federation (2013-04-16)
V Murali, Soon-An Ong, Li-Ngee Ho, Yee-Shian Wong, Nasrul Hamidin
ABSTRACT
Microbial fuel cells (MFCs) represent an emerging technology that focuses on power generation and effluent treatment. This review compiles articles related to MFCs using azo dye as the substrate. The significance of the general components in MFCs and systems of MFCs treating azo dye is depicted in this review. In addition, degradation of azo dyes such as Congo red, methyl orange, active brilliant red X-3B, amaranth, reactive blue 221, and acid orange 7 in MFCs are summarized. Further exploration and operational modification are suggested to address the challenges of complete removal of azo dye with maximum power generation in an MFC. In addition, a sequential treatment system with MFCs is suggested for complete mineralization of azo dye.
Product Description
Supelco
Amaranth, analytical standard
Sigma-Aldrich
Methyl Orange, suitable for microscopy
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Amaranth, Dye content 85-95 %