Merck
  • Physiological roles of mycothiol in detoxification and tolerance to multiple poisonous chemicals in Corynebacterium glutamicum.
Physiological roles of mycothiol in detoxification and tolerance to multiple poisonous chemicals in Corynebacterium glutamicum.
Archives of microbiology (2013-04-26)
Ying-Bao Liu, Ming-Xiu Long, Ya-Jie Yin, Mei-Ru Si, Lei Zhang, Zhi-Qiang Lu, Yao Wang, Xi-Hui Shen
ABSTRACT
Mycothiol (MSH) plays important roles in maintaining cytosolic redox homeostasis and in adapting to reactive oxygen species in the high-(G + C)-content Gram-positive Actinobacteria. However, its physiological roles are ill defined compared to glutathione, the functional analog of MSH in Gram-negative bacteria and most eukaryotes. In this research, we explored the impact of intracellular MSH on cellular physiology by using MSH-deficient mutants in the model organism Corynebacterium glutamicum. We found that intracellular MSH contributes significantly to resistance to alkylating agents, glyphosate, ethanol, antibiotics, heavy metals and aromatic compounds. In addition, intracellular MSH is beneficial for withstanding oxidative stress induced by various oxidants in C. glutamicum. This study greatly expanded our current knowledge on the physiological functions of mycothiol in C. glutamicum and could be applied to improve the robustness of this scientifically and commercially important species in the future.
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Supelco
Resorcinol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resorcinol, ≥98%, FG
Supelco
Melting point standard 79-81°C, analytical standard