Merck
  • The N-terminal region of two-pore channel 1 regulates trafficking and activation by NAADP.
The N-terminal region of two-pore channel 1 regulates trafficking and activation by NAADP.
The Biochemical journal (2013-05-03)
Dev Churamani, Robert Hooper, Taufiq Rahman, Eugen Brailoiu, Sandip Patel
ABSTRACT
TPCs (two-pore channels) are NAADP (nicotinic acid-adenine dinucleotide phosphate)-sensitive Ca2+-permeable ion channels expressed on acidic organelles. In the present study we show that deletion of the N-terminal region redirects TPC1 to the ER (endoplasmic reticulum). The introduction of fluorophores at the N-terminus of TPC1 does not affect its subcellular location, but does reversibly abolish NAADP sensitivity. Our results reveal a dual role for the N-terminus in localization and function of TPC1.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
Nicotinic acid adenine dinucleotide phosphate sodium salt, ≥92.5%