Merck
  • Histone deacetylases inhibitor trichostatin A modulates the extracellular release of APE1/Ref-1.
Histone deacetylases inhibitor trichostatin A modulates the extracellular release of APE1/Ref-1.
Biochemical and biophysical research communications (2013-05-15)
Sunga Choi, Yu Ran Lee, Myoung Soo Park, Hee Kyoung Joo, Eun Jung Cho, Hyo Shin Kim, Cuk Seong Kim, Jin Bong Park, Kaikobad Irani, Byeong Hwa Jeon
ABSTRACT
Apurinic/apyrimidinic endonuclease 1/Redox factor-1 (APE1/Ref-1) can be acetylated via post-translational modification. We investigated the effect of an inhibitor of histone deacetylases on the extracellular release of APE1/Ref-1 in HEK293 cells. Trichostatin A (TSA), an inhibitor of histone deacetylases, induced APE1/Ref-1 secretion without changing cell viability. In a fluorescence quantitative assay, the secreted APE1/Ref-1 was estimated to be about 10 ng/mL in response to TSA (1 μM). However, TSA did not induce the secretion of lysine-mutated APE1/Ref-1 (K6R/K7R). TSA also caused nuclear to cytoplasmic translocation of APE1/Ref-1. Taken together, these findings suggest that APE1/Ref-1 is a protein whose secretion is governed by lysine acetylation.
Product Description
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)