Merck
  • Chemical composition and anti-Trypanosoma cruzi activity of essential oils obtained from leaves of Xylopia frutescens and X. laevigata (Annonaceae).
Chemical composition and anti-Trypanosoma cruzi activity of essential oils obtained from leaves of Xylopia frutescens and X. laevigata (Annonaceae).
Natural product communications (2013-05-18)
Thanany Brasil da Silva, Leociley Rocha Alencar Menezes, Marília Fernanda Chaves Sampaio, Cássio Santana Meira, Elisalva Teixeira Guimarães, Milena Botelho Pereira Soares, Ana Paula do Nascimento Prata, Paulo Cesar de Lima Nogueira, Emmanoel Vilaça Costa
ABSTRACT
Essential oils from leaves of Xylopia frutescens (XFMJ) and two specimens of Xylopia laevigata (XLMC and XLSI) were obtained by hydrodistillation using a Clevenger-type apparatus, and analyzed by GC-MS and GC-FID. Sesquiterpenes dominated the essential oils. The main constituents of XFMJ were (E)-caryophyllene (24.8%), bicyclogermacrene (20.8%), germacrene D (17.0%), beta-elemene (7.9%), and (E)-beta-ocimene (6.8%). XLMC contained significant quantities of germacrene D (18.9%), bicyclogermacrene (18.4%), beta-elemene (9.5%), delta-selinene (9.2%), (E)-caryophyllene (8.5%), germacrene B (5.7%) and gamma-muurolene (5.7%), while germacrene D (27.0%), bicyclogermacrene (12.8%), (E)-caryophyllene (8.6%), gamma-muurolene (8.6%), delta-cadinene (6.8%), and germacrene B (6.0%) were the main components of XLSI. The essential oils had trypanocidal activity against the Y strain of Trypanosoma cruzi, with IC50 values lower than 30 microg x mL(-1) and 15 microg x mL(-1) against epimastigote and trypomastigote forms of T. cruzi, respectively, and were also able to reduce the percentage in vitro of T. cruzi-infected macrophages and the intracellular number of amastigotes at concentrations that were non-cytotoxic to macrophages.
Product Description
Supelco
(−)-trans-Caryophyllene, analytical standard
Sigma-Aldrich
(−)-trans-Caryophyllene, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
Ocimene, mixture of isomers, stabilized, ≥90%
Sigma-Aldrich
β-Caryophyllene, ≥80%, FCC, FG