Merck
  • Topical and systemic therapies for oral and perioral herpes simplex virus infections.
Topical and systemic therapies for oral and perioral herpes simplex virus infections.
Journal of the California Dental Association (2013-05-28)
Eric T Stoopler, Ramesh Balasubramaniam
ABSTRACT
Oral and perioral herpes simplex virus (HSV) infections in healthy individuals often present with signs and symptoms that are clearly recognized by oral health care providers (OHCPs). Management of these infections is dependent upon a variety of factors and several agents may be used for treatment to accelerate healing and decrease symptoms associated with lesions. This article will review the pertinent aspects of topical and systemic therapies of HSV infections for the OHCP.
Product Description
Aciclovir, European Pharmacopoeia (EP) Reference Standard
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Aminopurine, ≥99%
SAFC
L-Valine
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
Sigma-Aldrich
Valacyclovir hydrochloride hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Famciclovir, ≥98% (HPLC)