Merck
  • A pilot study of salvage irinotecan monotherapy for advanced biliary tract cancer.
A pilot study of salvage irinotecan monotherapy for advanced biliary tract cancer.
Anticancer research (2013-06-12)
Takashi Sasaki, Hiroyuki Isayama, Yousuke Nakai, Naminatsu Takahara, Yumiko Satoh, Daiya Takai, Hirofumi Kogure, Natsuyo Yamamoto, Kenji Hirano, Minoru Tada, Yutaka Yatomi, Kazuhiko Koike
ABSTRACT
To evaluate the treatment outcomes of irinotecan monotherapy for patients with advanced biliary tract cancer refractory to gemcitabine, cisplatin, and oral fluoropyrimidine. Irinotecan (100 mg/m(2)) was administered intravenously on days 1, 8, and 15, repeated every four weeks. Thirteen patients were enrolled. The dose intensity was only 55.0%. The response rate and disease control rate were 1/13 (7.7%) and 3/13 (23.1%), respectively. The median overall survival and time-to-progression were 6.7 months (95% confidence interval=3.0-10.4 months) and 1.8 months (95% confidence interval=1.6-3.9 months), respectively. Grade 3/4 adverse events included leukopenia (7/13), neutropenia (8/13), anemia (6/13), nausea (1/13), vomiting (1/13), anorexia (2/13), diarrhea (1/13), and constipation (1/13). Irinotecan monotherapy had a modest antitumor effect even for patients who were refractory to gemcitabine, cisplatin, and oral fluoropyrimidine. However, this regimen was not fully tolerated as third-line or fourth-line therapy. Therefore, further evaluation of a modified irinotecan regimen is necessary.
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE