Merck
  • Electrochemical properties of Sn-substituted LiMn2O4 thin films prepared by radio-frequency magnetron sputtering.
Electrochemical properties of Sn-substituted LiMn2O4 thin films prepared by radio-frequency magnetron sputtering.
Journal of nanoscience and nanotechnology (2013-07-19)
Woo Yeon Kong, Haena Yim, Seok-Jin Yoon, Sahn Nahm, Ji-Won Choi
ABSTRACT
The LiMn2O4 and LiSn0.0125Mn1975O4 thin films were grown on Pt/Ti/SiO2/Si (100) substrate by RF magnetron sputtering. To obtain the structural stability and good cycle performance, deposition parameters, namely working pressure, sputtering gas ratio of Ar and O2, post-annealing temperature were established. The structure and surface morphology of thin films were characterized by X-ray diffraction (XRD) and scanning electron microscopy (SEM), respectively. The electrochemical properties were estimated by two electrode half-cell test with WBCS 3000 (Wonatech, Korea) at constant current rate of 1 C-rate. The Sn substituted LiMn2O4 thin film deposited at 10 mtorr with mixture of argon and oxygen (Ar/O2 = 3/1) and then annealed at 500 degrees C in O2 atmosphere showed good cycle performance. The Sn substituted LiMn2O4 thin films showed larger capacity of -30 microAh/microm-cm2 and higher cyclability than LiMn2O4 thin films.
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis