Merck
  • Studies on antifungal properties of phenol complexes with secondary amines. 2. Antifungal properties in vitro of the complex of 2,4,5-trichlorophenol with dicyclohexylamine.
Studies on antifungal properties of phenol complexes with secondary amines. 2. Antifungal properties in vitro of the complex of 2,4,5-trichlorophenol with dicyclohexylamine.
Mycoses (1989-07-01)
R Witek, W Witek, B Walów, S J Pietr, A Kubis, Z Jopek
ABSTRACT
In studies in vitro it has been found that 2,4,5-trichlorophenol in the form of a complex with dicyclohexylamine is two times more active an antifungal agent than free chlorophenol.
Sigma-Aldrich
Dicyclohexylamine, 99%
Sigma-Aldrich
Dicyclohexylamine nitrite, 97%