Merck
  • An antioxidant induced alterations in peroxidase activity in ageing Zaprionus paravittiger (Diptera).
An antioxidant induced alterations in peroxidase activity in ageing Zaprionus paravittiger (Diptera).
Mechanisms of ageing and development (1988-11-30)
R Wadhwa, M Kaur, S P Sharma
ABSTRACT
Sodium hypophosphite (1 x 10(3) microM) supplementation in the diet of Zaprionus paravittiger resulted in adaptive responses in the quantitative as well as qualitative activity of peroxidase. The changes in peroxidase were in compliance with the earlier reported sodium hypophosphite (SHP)-induced prolonged life span of flies (Wadhwa, R., Arch. Gerontol. Geriatr., 6 (1987) 141-147) and support the concept that antioxidative enzymes may serve as important longevity determinants.
Sigma-Aldrich
Sodium hypophosphite, puriss., meets analytical specification of BPC63, anhydrous, 98-101%
Sigma-Aldrich
Sodium hypophosphite monohydrate, ≥99%