Merck
  • Phosphines as a new structural probe of hemoglobin. 1H-NMR evidence for perturbations in the beta heme pocket induced by a thiol reagent.
Phosphines as a new structural probe of hemoglobin. 1H-NMR evidence for perturbations in the beta heme pocket induced by a thiol reagent.
Biochimica et biophysica acta (1987-08-21)
A Bondon, P Sodano, G Simonneaux, C T Craescu
ABSTRACT
Binding of trimethylphosphine to myoglobins and hemoglobins from a variety of sources has been examined by 1H-nuclear magnetic resonance. The hemoglobins exhibit two resonances at high field (approx. -3.5 ppm) which have been assigned to PMe3 bound to alpha or to beta subunits. Perturbations in the beta heme pocket induced by a thiol reagent have been detected both in 1H and 31P spectra.
Sigma-Aldrich
Trimethylphosphine, 97%
Sigma-Aldrich
Trimethylphosphine solution, 1.0 M in THF
Sigma-Aldrich
Trimethylphosphine solution, 1.0 M in toluene